Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Horizon™ CD105 Mouse, anti-Human, RY586, Clone: 266, BD Horizon™

Catalog No. BDB568141 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB568141 100 Tests
BDB568142 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. BDB568141 Supplier BD Horizon™ Supplier No. 568141
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

The 266 monoclonal antibody specifically binds to CD105. CD105 is a type I transmembrane glycoprotein that is encoded by END (Endoglin) and belongs to the transforming growth factor-β (TGF-β) type III receptor family. CD105 is expressed on cells as a homodimer comprised of ∼95 kDa subunits. CD105 is expressed on vascular endothelial cells and placental syncytiotrophoblasts and at lower levels on stromal fibroblasts. It is also expressed on mesenchymal stem cells, erythroid precursors, activated macrophages, pre-B cells, and some tumor cells and cell lines including U937 cells. CD105 serves as a regulatory component of the TGF-β receptor system. In association with TGF- βRI or TGF- βRII, CD105 binds TGF-β1 and TGF-β3 with high affinity but does not bind to TGF-β2. Expression of CD105 is increased on activated endothelium in tissues undergoing angiogenesis, such as in tumors, or in cases of wound healing or dermal inflammation.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD105
Applications Flow Cytometry
Clone 266
Conjugate RY586
Host Species Mouse
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.