Learn More
Invitrogen™ CD105 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA595428
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human SiHa whole cell, human Hela whole cell. Flow: U87 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
CD105 or Endoglin is a Type I transmembrane protein, which is highly expressed on human vascular endothelial cells. It exists on an O- and N-glycosylated homodimer. Up regulation of endoglin expression has been demonstrated in tumor vasculature and proliferating cells, suggesting that it is a proliferation associated endothelial marker. CD105 binds to TGF beta 1 and 3 with high affinity but not to TGF beta 2.
Specifications
CD105 | |
Polyclonal | |
Unconjugated | |
ENG | |
AI528660; AI662476; CD105; CD105 antigen; cell surface MJ7/18 antigen; Edg; END; Endo; Endoglin; endoglin (Osler-Rendu-Weber syndrome 1); Eng; HHT1; ORW; ORW1; S-endoglin; transmembrane glycoprotein | |
Rabbit | |
Affinity chromatography | |
RUO | |
2022 | |
-20°C | |
Lyophilized |
Flow Cytometry, Western Blot | |
500 μg/mL | |
PBS with 4mg trehalose and no preservative | |
P17813 | |
ENG | |
A synthetic peptide corresponding to a sequence in the middle region of human CD105 (258-297aa YVSWLIDANHNMQIWTTGEYSFKIFPEKNIRGFKLPDTPQ). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.