Learn More
Invitrogen™ CD105 Recombinant Rabbit Monoclonal Antibody (103)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA529234
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human Endoglin/CD105.
CD105 or Endoglin is a Type I transmembrane protein, which is highly expressed on human vascular endothelial cells. It exists on an O- and N-glycosylated homodimer. Up regulation of endoglin expression has been demonstrated in tumor vasculature and proliferating cells, suggesting that it is a proliferation associated endothelial marker. CD105 binds to TGF beta 1 and 3 with high affinity but not to TGF beta 2.
Specifications
CD105 | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
P17813 | |
ENG | |
Recombinant Human Endoglin/CD105 protein (Met1-Gly586). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Flow Cytometry, Immunocytochemistry | |
103 | |
Unconjugated | |
ENG | |
AI528660; AI662476; CD105; CD105 antigen; cell surface MJ7/18 antigen; Edg; END; Endo; Endoglin; endoglin (Osler-Rendu-Weber syndrome 1); Eng; HHT1; ORW; ORW1; S-endoglin; transmembrane glycoprotein | |
Rabbit | |
Protein A | |
RUO | |
2022 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.