Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD106 Mouse anti-Human, Brilliant Violet 786, Clone: 51-10C9, BD Optibuild™

Catalog No. BDB744313 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB744313 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB744313 Supplier BD Biosciences Supplier No. 744313
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 51-10C9 monoclonal antibody specifically binds to CD106. CD106 is a 100-110 kDa type I transmembrane sialoglycoprotrein that is also known as Vascular cell adhesion molecule-1 (VCAM-1) and INCAM-110. CD106 is expressed at high levels on the surface of cytokine-stimulated endothelium, and at minimal levels on unstimulated endothelium. VCAM-1 serves as a ligand for the leukocyte integrinsα4β1 (CD49d/CD29 complex; VLA-4) andα4β7 (LPAM-1). The 51-10C9 monoclonal antibody inhibits the in vitro binding of lymphocytes and monocytes to VCAM-1 on stimulated endothelium.

The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 786nm. BD Horizon BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60nm filter). BV786 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 786 nm. BV786 offers a bright choice for the 6th detector off the violet laser.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD106
Classification Monoclonal
Clone 51-10C9
Concentration 0.2mg/mL
Conjugate Brilliant Violet 786
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias VCAM-1, Vascular cell adhesion protein 1, INCAM-100, L1CAM
Host Species Mouse
Immunogen Human VCAM-1 Recombinant Protein
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.