Learn More
CD106 Mouse anti-Human, PE, Clone: 51-10C9, BD
Mouse Monoclonal Antibody
$132.00 - $294.00
Specifications
Antigen | CD106 |
---|---|
Clone | 51-10C9 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | PE |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
BDB561679
|
BD Biosciences
561679 |
25 Tests |
Each for $132.00
|
|
|||||
BDB555647
|
BD Biosciences
555647 |
100 Tests |
Each for $294.00
|
|
|||||
Description
The 51-10C9 monoclonal antibody specifically binds to CD106. CD106 is a 100-110 kDa type I transmembrane sialoglycoprotrein that is also known as Vascular cell adhesion molecule-1 (VCAM-1) and INCAM-110. CD106 is expressed at high levels on the surface of cytokine-stimulated endothelium, and at minimal levels on unstimulated endothelium. VCAM-1 serves as a ligand for the leukocyte integrins α4β1 (CD49d/CD29 complex; VLA-4) and α4β7 (LPAM-1). The 51-10C9 monoclonal antibody inhibits the in vitro binding of lymphocytes and monocytes to VCAM-1 on stimulated endothelium.
Specifications
CD106 | |
Flow Cytometry | |
PE | |
RUO | |
Human | |
IgG1 κ | |
Affinity Purified | |
VCAM-1; Vascular cell adhesion protein 1; INCAM-100; L1CAM |
51-10C9 | |
Monoclonal | |
Mouse | |
Immunology | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.