Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD107a Mouse anti-Human , Alexa Fluor 488, Clone: H4A3, BD Biosciences™
Mouse Monoclonal Antibody
$132.60 - $313.50
Specifications
Antigen | CD107a |
---|---|
Clone | H4A3 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Alexa Fluor 488 |
Description
CD107a Monoclonal antibody specifically detects CD107a in Human samples. It is validated for Flow CytometrySpecifications
CD107a | |
Flow Cytometry | |
Alexa Fluor 488 | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human Adult Adherent Peripheral Blood Cells | |
Primary |
H4A3 | |
Monoclonal | |
Mouse | |
Human | |
LAMP1; LAMP-1; LAMPA; LGP120 | |
IgG1 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to Alexa Fluor 488 under optimum conditions, and unreacted Alexa Fluor 488 was removed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title