Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD107a Mouse anti-Human, CoraLite 647, Clone: H4A3, Proteintech
SDP

Catalog No. 502258020 Shop All Proteintech Group Inc Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Conjugate:
CoraLite 647
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Conjugate
50-225-8020 CoraLite 647
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-225-8020 Supplier Proteintech Group Inc Supplier No. CL64765051100TESTS
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

The CD107a antibody from Proteintech is a mouse monoclonal antibody to a human adherent peripheral blood cells of human CD107a. This antibody recognizes human antigen. The CD107a antibody has been validated for the following applications: IF, FC analysis. This antibody is coralite™647.

Specifications

Antigen CD107a
Applications Immunofluorescence, Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone H4A3
Concentration 5 μL/Test
Conjugate CoraLite 647
Host Species Mouse
Primary or Secondary Primary
Gene ID (Entrez) 3916
Target Species Human
Isotype IgG1
No. of Reactions 100 Tests
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.