Learn More
Invitrogen™ CD107b (LAMP-2) Monoclonal Antibody (M3/84), FITC, eBioscience™

Description
The M3/84 monoclonal antibody reacts with mouse Mac-3, the 110 kDa protein expressed by mononuclear phagocytes. The Mac-3 antigen is expressed intracellularly by bone marrow monocyte lineage and is upregulated during macrophage differentiation. Peritoneal and tissue macrophages, dendritic cells, and endothelial cells express this antigen on their surface. It is reported that Mac-3 may be identical to CD107b. Applications Reported: The M3/84 antibody has been reported for use in flow cytometric analysis. Applications Tested: The M3/84 antibody has been tested by flow cytometric analysis of mouse bone marrow and thioglycolate-elicited peritoneal exudates cells and can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
Specifications
Specifications
| Antigen | CD107b (LAMP-2) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | M3/84 |
| Concentration | 0.5 mg/mL |
| Conjugate | FITC |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Gene | LAMP2 |
| Gene Accession No. | P17047 |
| Gene Alias | CD107 antigen-like family member B; CD107b; EGK_20887; Igp96; Lamp II; LAMP2; LAMP-2; LAMP-2 Isoform 2A; LAMP-2 Variant A; Lamp-2a; Lamp-2b; Lamp-2c; LAMPB; LGP110; LGP-110; LGP-96; LGP-B; lysosomal associated membrane protein 2; lysosomal membrane glycoprotein 2; lysosomal membrane glycoprotein type B; lysosomal-associated membrane protein 2; lysosome-associated membrane glycoprotein 2; lysosome-associated membrane protein 2; Mac3; RP23-193O17.2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.