Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD109 Antibody (496920), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB4385MFV500
Description
CD109 Monoclonal antibody specifically detects CD109 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOFSpecifications
| CD109 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry, CyTOF | |
| 496920 | |
| Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
| 150 kDa TGF-beta-1-binding protein, activated T-cell marker CD109, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 7, CD109 antigen, CD109 antigen (Gov platelet alloantigens), CD109 molecule, CPAMD7r150, DKFZp762L1111, FLJ38569, FLJ41966, Gov platelet alloantigens, p180, Platelet-specific Gov antigen, RP11-525G3.1 | |
| Mouse myeloma cell line NS0-derived recombinant human CD109, Val22-Ser1268 (Tyr703Ser and Thr1241Met), Accession # Q6YHK3 | |
| 0.1 mL | |
| Cancer, Cell Biology, Cytokine Research, Immunology, Stem Cell Markers | |
| 135228 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction