Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD109 Antibody (B-E47), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP318101AF405
Description
CD109 Monoclonal antibody specifically detects CD109 in Human samples. It is validated for Flow CytometrySpecifications
CD109 | |
Monoclonal | |
Alexa Fluor 405 | |
150 kDa TGF-beta-1-binding protein, activated T-cell marker CD109, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 7, CD109 antigen, CD109 antigen (Gov platelet alloantigens), CD109 molecule, CPAMD7r150, DKFZp762L1111, FLJ38569, FLJ41966, Gov platelet alloantigens, p180, Platelet-specific Gov antigen, RP11-525G3.1 | |
Eahy 926 cell line. Hybridoma: Myeloma X63/AG.8653 x Balb/c spleen cells | |
0.1 mL | |
Cancer, Cell Biology, Cytokine Research, Immunology, Stem Cell Markers | |
135228 | |
Store at 4°C in the dark. | |
IgG1 κ |
Flow Cytometry | |
B-E47 | |
50mM Sodium Borate | |
Mouse | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction