Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD109 Antibody (W7C5), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP275969AF700
Description
CD109 Monoclonal specifically detects CD109 in Human samples. It is validated for Flow Cytometry, Flow (Cell Surface).Specifications
CD109 | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CD109 | |
WERI-RB-1 retinoblastoma cell line | |
0.1 mL | |
Primary | |
The antibody MEM-174 recognizes CD11b antigen (Mac-1 alpha), a 165-170 kDa type I transmembrane protein mainly expressed on monocytes, granulocytes and NK-cells. The CD11b mediates neutrophil and monocyte interactions with stimulated endothelium. | |
Store at 4°C in the dark. |
Flow Cytometry | |
W7C5 | |
Flow Cytometry, Flow (Cell Surface) | |
150 kDa TGF-beta-1-binding protein, activated T-cell marker CD109, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 7, CD109 antigen, CD109 antigen (Gov platelet alloantigens), CD109 molecule, CPAMD7r150, DKFZp762L1111, FLJ38569, FLJ41966, Gov platelet alloantigens, p180, Platelet-specific Gov antigen, RP11-525G3.1 | |
Mouse | |
Protein A purified | |
Cancer, Cell Biology, Cytokine Research, Immunology, Stem Cell Markers | |
135228 | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction