Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD109 Monoclonal Antibody (496920)
GREENER_CHOICE

Catalog No. PIMA524208
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524208 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524208 Supplier Invitrogen™ Supplier No. MA524208
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute at 0.5 mg/mL in sterile PBS.

CD109 is a member of the alpha2-macroglobulin/complement superfamily. The encoded GPI-linked glycoprotein is found on the cell surface of platelets, activated T-cells, and endothelial cells. The protein binds to and negatively regulates signaling of transforming growth factor beta (TGF-beta).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD109
Applications Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 496920
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene CD109
Gene Accession No. Q6YHK3
Gene Alias 150 kDa TGF-beta-1-binding protein; 9930012E15Rik; activated T-cell marker CD109; AI480638; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 7; Cd109; CD109 antigen; CD109 molecule; CPAMD7; GARP; Gov platelet alloantigens; GPI-anchored alpha 2 macroglobulin-related protein; GPI-anchored alpha-2 macroglobulin-related protein; p180; platelet-specific Gov antigen; r150; RP11-525G3.1
Gene Symbols CD109
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human CD109 Val22-Ser1268 (Tyr703Ser and Thr1241Met).
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 135228
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.