Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD109 Mouse anti-Human, BUV395, Clone: TEA 2/16, BD Optibuild™

Catalog No. BDB744496 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB744496 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB744496 Supplier BD Biosciences Supplier No. 744496
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The TEA 2/16 monoclonal antibody specifically recognizes CD109 which is also known as C3 and PZP-like alpha-2-macroglobulin domain-containing protein 7 (CPAMD7), Gov platelet alloantigens, or 150 kDa TGF-beta-1-binding protein. CD109 is a 150-170 kDa glycosylphosphatidylinositol (GPI)-linked glycoprotein expressed on activated T cells, myeloid progenitor (CD34+) and mature myeloid lineage cells (monocytes, granulocytes, platelets), but not on CD34+ lymphoid progenitor cells. CD109 is also expressed on vein and artery endothelial cells. The expression of CD109 is upregulated on PHA-stimulated T cells. CD109 is reportedly expressed on long-term adult bone marrow cultured cells, where it is co-expressed with CD34 and CD90. Its biological role in hematopoiesis has not been fully elucidated. It may play a role in the negative regulation of transforming growth factor beta receptor signaling.

The antibody was conjugated to BD Horizon™ BUV395 which is part of the BD Horizon Brilliant™ Ultraviolet family of dyes. This dye has been exclusively developed by BD Biosciences to have minimal spillover into other detectors, making it an optimal choice for multicolor flow cytometry. With an Ex Max at 348 nm and an Em Max at 395 nm, BD Horizon BUV395 can be excited with a 355 nm laser and detected with a 379/28 filter. BUV395 is a UV-excitable dye that has been developed exclusively by BD Biosciences. With an excitation max of 348 nm and emission max of 395 nm, BUV395 can be excited by the 355nm laser and detected in a 379/28 filter. This dye is optimal for multicolor flow cytometry because it has little to no spillover into other detectors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD109
Classification Monoclonal
Clone TEA 2/16
Concentration 0.2mg/mL
Conjugate Brilliant Ultraviolet 395
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias CPAMD7, 8A3, E123, 7D1, p180, r150, Gov platelet alloantigens
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.