Learn More
CD109 Mouse anti-Human, Clone: 2H8G7, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | CD109 |
---|---|
Clone | 2H8G7 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6283
![]() ![]() |
Proteintech Group Inc
601791IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
There are 2 forms of 150 kDa (p150) and 120 kDa (p120) due to proteolytic degradation from a 180 kDa form. The antibody can recognize all isoforms.
CD109 is a member of the alpha2-macroglobulin/complement superfamily. The encoded GPI-linked glycoprotein is found on the cell surface of platelets, activated T-cells, and endothelial cells. The protein binds to and negatively regulates signaling of transforming growth factor beta (TGF-beta).Specifications
CD109 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
CD109, CD109 antigen, CD109 molecule, CPAMD7, p180, Platelet specific Gov antigen, r150, RP11 525G3.1 | |
CD109 | |
IgG1 | |
Caprylic Acid Precipitation | |
Antibody |
2H8G7 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q6YHK3 | |
135228 | |
Peptide | |
Primary | |
-20°C | |
CD109 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.