Learn More
Invitrogen™ CD109 Polyclonal Antibody

Description
CD109 Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | CD109 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | CD109 |
| Gene Accession No. | Q6YHK3, Q8R422 |
| Gene Alias | 150 kDa TGF-beta-1-binding protein; 9930012E15Rik; activated T-cell marker CD109; AI480638; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 7; Cd109; CD109 antigen; CD109 molecule; CPAMD7; GARP; Gov platelet alloantigens; GPI-anchored alpha 2 macroglobulin-related protein; GPI-anchored alpha-2 macroglobulin-related protein; p180; platelet-specific Gov antigen; r150; RP11-525G3.1 |
| Gene Symbols | CD109 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.