Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD109 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$378.00 - $607.50
Specifications
Antigen | CD109 |
---|---|
Dilution | Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml |
Applications | Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
CD109 Polyclonal antibody specifically detects CD109 in Human samples. It is validated for ImmunofluorescenceSpecifications
CD109 | |
Immunofluorescence | |
Unconjugated | |
Rabbit | |
Cancer, Cell Biology, Cytokine Research, Immunology, Stem Cell Markers | |
PBS, pH 7.2, 40% glycerol | |
135228 | |
IgG | |
Affinity purified |
Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Human | |
150 kDa TGF-beta-1-binding protein, activated T-cell marker CD109, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 7, CD109 antigen, CD109 antigen (Gov platelet alloantigens), CD109 molecule, CPAMD7r150, DKFZp762L1111, FLJ38569, FLJ41966, Gov platelet alloantigens, p180, Platelet-specific Gov antigen, RP11-525G3.1 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: VVSGNKRLKELSYMVVSRGQLVAVGKQNSTMFSLTPENSWTPKACVIVYYIEDDGEIISDVLKIPVQLVFKNKIKLYWSKVKAEPSEKVSLRISVTQPDSIVGIVAVDKSVNLMNASNDITMENVVHEL | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title