Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD110 Mouse anti-Human, Brilliant Violet 650, Clone: 1.6.1, BD Optibuild™

Catalog No. BDB743579 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB743579 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB743579 Supplier BD Biosciences Supplier No. 743579
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 1.6.1 monoclonal antibody specifically binds to the human Thrombopoietin Receptor (TPO-R) that is also known as the Myeloproliferative leukemia protein (c-Mpl) or CD110. CD110 is a type I transmembrane glycoprotein and a member of the hematopoietin receptor family. It is expressed on hematopoietic stem cells, a subfraction of hematopoietic precursor cells, cells of the megakaryocytic lineage and platelets. CD110 serves as a receptor for thrombopoietin. Upon binding of thrombopoietin to CD110, megakaryocyte proliferation and differentiation is induced, platelets are produced and stem cells are protected from apoptosis.

The antibody was conjugated to BD Horizon™ BV650 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 650nm. BD Horizon BV650 can be excited by the violet laser and detected in a filter used to detect APC-like dyes (eg, 660/20nm filter). Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. However, the spillover can be corrected through compensation as with any other dye combination. BV650 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 650 nm. Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. BV650 will have moderate spillover into the BD Horizon™ BV711 detector.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD110
Classification Monoclonal
Clone 1.6.1
Concentration 0.2mg/mL
Conjugate Brilliant Violet 650
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias C-MPL, MPL, MPLV, TPOR, Thrombopoietin receptor
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.