Learn More
Invitrogen™ CD112 (Nectin-2) Monoclonal Antibody (R2.477), APC, eBioscience™
Mouse Monoclonal Antibody
$408.00
Specifications
Antigen | CD112 (Nectin-2) |
---|---|
Clone | R2.477 |
Concentration | 5 μL/Test |
Content And Storage | 4°C, store in dark, DO NOT FREEZE! |
Applications | Flow Cytometry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-112-3070
![]() |
Invitrogen™
17112842 |
100 Tests |
Each of 1 for $408.00
|
![]() |
|||||
Description
Description: This R2.477 monoclonal antibody reacts with human CD112 (also known as Nectin-2 or Poliovirus Receptor Related 2), a 60-65 kDa immunoglobulin-like transmembrane cell adhesion molecule. This cell surface molecule is expressed on hematopoietic cells, endothelial, epithelial, as well as tumor cell lines such as Jurkat, U937, and MCF7. CD112 is a ligand for DNAM-1 (CD226), a leukocyte adhesion molecule expressed on natural killer (NK) cells. Interaction between CD112 and DNAM-1 promotes NK cytotoxic activity, leading to tumor cell lysis. In addition to its role in tumor suppression, CD112 plays a cooperative role in cell-cell adhesion, particularly in the formation of adherens junctions between neighboring epithelial cells and fibroblasts. CD112 can form homophilic and heterophilic interactions to regulate contact inhibition of cell movement and proliferation. Finally, CD112 has been reported to be a receptor for herpesvirus entry into cells. This monoclonal antibody has been reported to block cell adhesion and herpesvirus entry. Applications Reported: This R2.477 antibody has been reported for use in flow cytometric analysis. Applications Tested: This R2.477 antibody has been pre-titrated and tested by flow cytometric analysis of human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirical...
This gene encodes a single-pass type I membrane glycoprotein with two Ig-like C2-type domains and an Ig-like V-type domain. This protein is one of the plasma membrane components of adherens junctions. It also serves as an entry for certain mutant strains of herpes simplex virus and pseudorabies virus, and it is involved in cell to cell spreading of these viruses. Variations in this gene have been associated with differences in the severity of multiple sclerosis. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.Specifications
CD112 (Nectin-2) | |
5 μL/Test | |
Flow Cytometry | |
APC | |
Mouse | |
RUO | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
Q92692 | |
5819 | |
Primary | |
Affinity Chromatography |
R2.477 | |
4°C, store in dark, DO NOT FREEZE! | |
Monoclonal | |
Liquid | |
IgG1 κ | |
Human | |
NECTIN2 | |
AI325026; AI987993; CD112; herpes virus entry mediator B; Herpesvirus entry mediator B; herpesvirus entry protein B; hveB; mHveB; MPH; Murine herpes virus entry protein B; murine herpesvirus entry protein B; nectin cell adhesion molecule 2; Nectin2; Nectin-2; Poliovirus receptor homolog; poliovirus receptor related 2; poliovirus receptor-like 2; poliovirus receptor-related 2; poliovirus receptor-related 2 (herpesvirus entry mediator B); poliovirus receptor-related protein 2; poliovirus sensitivity; PRR2; Pvr; Pvrl2; PVRR2; Pvs | |
NECTIN2 | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.