Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD112 (Nectin-2) Rat anti-Mouse, Brilliant Violet 421, Clone: 829038, BD OptiBuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 748046
Description
CD112 (Nectin-2) Monoclonal antibody specifically detects CD112 (Nectin-2) in Mouse samples. It is validated for Flow Cytometry.
Specifications
CD112 (Nectin-2) | |
Monoclonal | |
0.2 mg/mL | |
Rat | |
RUO | |
CD112 (Nectin-2) | |
Refrigerate (2 to 8°C) |
Flow Cytometry | |
829038 | |
Brilliant Violet 421 | |
50 μg | |
Primary | |
Mouse | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction