Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD112 (Nectin-2) Recombinant Rabbit Monoclonal Antibody (BLR071G)

Catalog No. PIMA544334
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544334 10 μL
PIMA544333 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA544334 Supplier Invitrogen™ Supplier No. MA544334
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Epitope retrieval with citrate buffer pH6.0 is recommended for FFPE cell sections.

This gene encodes a single-pass type I membrane glycoprotein with two Ig-like C2-type domains and an Ig-like V-type domain. This protein is one of the plasma membrane components of adherens junctions. It also serves as an entry for certain mutant strains of herpes simplex virus and pseudorabies virus, and it is involved in cell to cell spreading of these viruses. Variations in this gene have been associated with differences in the severity of multiple sclerosis. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD112 (Nectin-2)
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone BLR071G
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation BBS with 0.09% sodium azide; pH 7.8-8.6
Gene NECTIN2
Gene Accession No. Q92692
Gene Alias AI325026; AI987993; CD112; herpes virus entry mediator B; Herpesvirus entry mediator B; herpesvirus entry protein B; hveB; mHveB; MPH; Murine herpes virus entry protein B; murine herpesvirus entry protein B; nectin cell adhesion molecule 2; Nectin2; Nectin-2; Poliovirus receptor homolog; poliovirus receptor related 2; poliovirus receptor-like 2; poliovirus receptor-related 2; poliovirus receptor-related 2 (herpesvirus entry mediator B); poliovirus receptor-related protein 2; poliovirus sensitivity; PRR2; Pvr; Pvrl2; PVRR2; Pvs
Gene Symbols NECTIN2
Host Species Rabbit
Immunogen Peptide representing a region between residue 50 and 100 of human Nectin cell adhesion molecule 2.
Purification Method Purified
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5819
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.