Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD112 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984895
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4895 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4895 Supplier Bioss Supplier No. BS2679R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a single-pass type I membrane glycoprotein with two Ig-like C2-type domains and an Ig-like V-type domain. This protein is one of the plasma membrane components of adherens junctions. It also serves as an entry for certain mutant strains of herpes simplex virus and pseudorabies virus, and it is involved in cell to cell spreading of these viruses. Variations in this gene have been associated with differences in the severity of multiple sclerosis. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.

Specifications

Antigen CD112
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene NECTIN2
Gene Accession No. Q92692
Gene Alias AI325026; AI987993; CD112; Herpes virus entry mediator B; Herpesvirus entry mediator B; herpesvirus entry protein B; HVEB; mHveB; Mph; Murine herpes virus entry protein B; murine herpesvirus entry protein B; Nectin cell adhesion molecule 2; Nectin2; Nectin-2; Poliovirus receptor homolog; poliovirus receptor related 2; poliovirus receptor-like 2; poliovirus receptor-related 2; poliovirus receptor-related 2 (herpesvirus entry mediator B); Poliovirus receptor-related protein 2; poliovirus sensitivity; PRR2; Pvr; Pvrl2; PVRR2; Pvs
Gene Symbols NECTIN2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human nectin 2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5819
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.