Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD114 Mouse anti-Human, Brilliant Violet 786, Clone: LMM741, BD Optibuild™

Catalog No. BDB743523 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB743523 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB743523 Supplier BD Biosciences Supplier No. 743523
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The LMM741 monoclonal antibody specifically recognizes CD114 which is also known as the Granulocyte-Colony Stimulating Factor Receptor (G-CSFR). CD114 is a ñ150 kDa type I transmembrane glycoprotein that is encoded by CSF3R (colony stimulating factor 3 receptor) and belongs to the class 1 cytokine receptor family. CD114 is expressed on granulocytes, monocytes, dendritic cells, endothelial cells, platelets, placenta and myeloid leukemias and a variety of tumor cell lines. CD114 serves as the receptor for granulocyte colony stimulating factor (G-CSF) which plays a role in myeloid cell proliferation and differentiation. The immunogen used to generate this hybridoma was cells transfected with an expression vector containing a full-length cDNA encoding the human G-CSFR.

The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 786nm. BD Horizon BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60nm filter). BV786 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 786 nm. BV786 offers a bright choice for the 6th detector off the violet laser.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD114
Classification Monoclonal
Clone LMM741
Concentration 0.2mg/mL
Conjugate Brilliant Violet 786
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias GCSFR, G-CSF Receptor, G-CSF-R, G-CSFR, HG-CSFR, CSF3R
Host Species Mouse
Immunogen G-CSFR cDNA transfected cells.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.