Learn More
CD114 Mouse anti-Human, BUV395, Clone: LMM741, BD Optibuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 743524
Description
The LMM741 monoclonal antibody specifically recognizes CD114 which is also known as the Granulocyte-Colony Stimulating Factor Receptor (G-CSFR). CD114 is a ñ150 kDa type I transmembrane glycoprotein that is encoded by CSF3R (colony stimulating factor 3 receptor) and belongs to the class 1 cytokine receptor family. CD114 is expressed on granulocytes, monocytes, dendritic cells, endothelial cells, platelets, placenta and myeloid leukemias and a variety of tumor cell lines. CD114 serves as the receptor for granulocyte colony stimulating factor (G-CSF) which plays a role in myeloid cell proliferation and differentiation. The immunogen used to generate this hybridoma was cells transfected with an expression vector containing a full-length cDNA encoding the human G-CSFR.
The antibody was conjugated to BD Horizon™ BUV395 which is part of the BD Horizon Brilliant™ Ultraviolet family of dyes. This dye has been exclusively developed by BD Biosciences to have minimal spillover into other detectors, making it an optimal choice for multicolor flow cytometry. With an Ex Max at 348 nm and an Em Max at 395 nm, BD Horizon BUV395 can be excited with a 355 nm laser and detected with a 379/28 filter. BUV395 is a UV-excitable dye that has been developed exclusively by BD Biosciences. With an excitation max of 348 nm and emission max of 395 nm, BUV395 can be excited by the 355nm laser and detected in a 379/28 filter. This dye is optimal for multicolor flow cytometry because it has little to no spillover into other detectors.
Specifications
CD114 | |
LMM741 | |
Brilliant Ultraviolet 395 | |
GCSFR, G-CSF Receptor, G-CSF-R, G-CSFR, HG-CSFR, CSF3R | |
G-CSFR cDNA transfected cells. | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.