Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD114 Mouse, anti-Human, R718, Clone: LMM741, BD OptiBuild™

Catalog No. BDB751916 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB751916 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB751916 Supplier BD Biosciences Supplier No. 751916
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The LMM741 monoclonal antibody specifically recognizes CD114 which is also known as the Granulocyte-Colony Stimulating Factor Receptor (G-CSFR). CD114 is a ∼150 kDa type I transmembrane glycoprotein that is encoded by CSF3R (colony stimulating factor 3 receptor) and belongs to the class 1 cytokine receptor family. CD114 is expressed on granulocytes, monocytes, dendritic cells, endothelial cells, platelets, placenta and myeloid leukemias and a variety of tumor cell lines. CD114 serves as the receptor for granulocyte colony stimulating factor (G-CSF) which plays a role in myeloid cell proliferation and differentiation. The immunogen used to generate this hybridoma was cells transfected with an expression vector containing a full-length cDNA encoding the human G-CSFR.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD114
Applications Flow Cytometry
Classification Monoclonal
Clone LMM741
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.