Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD115 (c-fms) Monoclonal Antibody (04)
GREENER_CHOICE

Catalog No. PIMA535980
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535980 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535980 Supplier Invitrogen™ Supplier No. MA535980
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD115 (c-fms) Monoclonal Antibody for Flow

CSF1R (FMS) is involved in the production, differentiation, and function of macrophages, and is implicated in promyelocytic leukemias. Ligand binding activates the receptor kinase through a process of oligomerization and transphosphorylation. CSF1R is a tyrosine kinase transmembrane receptor and member of the CSF1/PDGF receptor family of tyrosine-protein kinases. Mutations in the gene encoding CSF1R have been associated with a predisposition to myeloid malignancy.Tyrosine-protein kinase that acts as cell-surface receptor for CSF1 and IL34 and plays an essential role in the regulation of survival, proliferation and differentiation of hematopoietic precursor cells, especially mononuclear phagocytes, such as macrophages and monocytes. Promotes the release of proinflammatory chemokines in response to IL34 and CSF1, and plays an important role in innate immunity and in inflammatory processes. The first intron of the CSF1R gene contains a transcriptionally inactive ribosomal protein L7 processed pseudogene oriented in the opposite direction. Mutations in the CSF1R gene have been associated with a predisposition to myeloid malignancy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD115 (c-fms)
Applications Flow Cytometry
Classification Monoclonal
Clone 04
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Csf1r
Gene Accession No. P07333
Gene Alias AI323359; CD115; CD115 antigen; C-FMS; colony stimulating factor 1 receptor; colony stimulating factor 1 receptor, formerly McDonough feline sarcoma viral (v-fms) oncogene homolog; CSF-1 receptor; CSF1R; CSF-1R; CSF-1-R; CSFIR; Csfmr; CSFR; EC 2.7.10.1; FIM2; Fim-2; FMS; fms proto-oncogene; HDLS; kinase CSFR; macrophage colony stimulating factor I receptor; macrophage colony-stimulating factor 1 receptor; McDonough feline sarcoma viral (v-fms) oncogene homolog; M-CSFR; M-CSF-R; mrfms; OTTHUMP00000224093; protein-tyrosine kinase; proto-oncogene c-Fms; proto-oncogene fms
Gene Symbols Csf1r
Host Species Mouse
Immunogen Recombinant Human CSF1R/MCSF Receptor/CD115 Protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1436
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.