Learn More
CD115 (CSF-1R) Rat anti-Mouse, Brilliant Violet 605, Clone: T38-320, BD Optibuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 743640
Description
The T38-320 monoclonal antibody specifically binds to CD115 which is also known as Colony stimulating factor 1 Receptor (CSF-1R) or Macrophage colony-stimulating factor 1 receptor (M-CSFR). This type I transmembrane glycoprotein is a receptor tyrosine kinase (RTK) that belongs to the Ig superfamily. It is expressed on a variety of cells including those committed to the mononuclear phagocyte lineage, such as, monocytes, macrophages, and osteoclasts. CSF-1 binds to and signals through CSF-1R homodimers which undergo tyrosine autophosphorylation and transduce downstream signaling pathways resulting in cytoskeletal reorganization and gene expression. CSF-1R activation stimulates the proliferation, differentiation, and survival of cells within the mononuclear phagocyte system. Interleukin-34 (IL-34) is another ligand for CD115 that can induce similar, as well as, some different biological responses by CD115-positive target cells.
This antibody is conjugated to BD Horizon™ BV605 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max of 602nm, BD Horizon BV605 can be excited by a violet laser and detected with a standard 610/20nm filter set. BD Horizon BV605 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an Em max at 605nm. Due to the excitation of the acceptor dye by the green (532 nm) and yellow-green (561 nm) lasers, there will be significant spillover into the PE and BD Horizon PE-CF594 detectors off the green or yellow-green lasers. BD Horizon BV605 conjugates are very bright, often exhibiting brightness equivalent to PE conjugates and can be used as a third color off of the violet laser. BV605 is a tandem fluorochrome that combines BD Horizon BV421 and Cy™3.5. These conjugates are very bright, exhibiting similar brightness to equivalent PE conjugates. Due to the excitation of Cy3.5 by the green (532nm) and yellow-green (561nm) lasers, there will be significant spillover of the BD Horizon BV605 signal into the PE and BD Horizon PE-CF594 detectors off the green or yellow-green lasers.
Specifications
CD115 (CSF-1R) | |
T38-320 | |
Brilliant Violet 605 | |
M-CSFR, M-CSF-R, CSF-1 Receptor, CSF-1R, Csf1r, Csfmr, Fms, c-Fms, Fim-2 | |
Mouse CD115 Recombinant Protein | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
Affinity Purified | |
RUO | |
Mouse | |
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.