Learn More
CD116 Monoclonal Antibody (4H1), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$1562.00 - $3905.00
Specifications
Antigen | CD116 |
---|---|
Clone | 4H1 |
Concentration | 0.5 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
14-116-937
![]() |
Invitrogen™
14116937 |
2 mg |
Each for $1,562.00
|
|
|||||
14-116-995
![]() |
Invitrogen™
14116995 |
10 mg |
Each for $3,905.00
|
|
|||||
Description
Description: The 4H1 monoclonal antibody reacts with the human CD116 molecule, the alpha subunit of GM-CSF receptor. The alpha subunit associates with the common beta chain (CD131) to form the high affinity receptor for GM-CSF. The GM-CSFR alpha chain is expressed by granulocytes, monocytes, endothelial cells, fibroblasts and some tumor cells. Applications Reported: 4H1 has been reported for use in flow cytometric analysis. Applications Tested: The 4H1 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.
The protein encoded by this gene is the alpha subunit of the heterodimeric receptor for colony stimulating factor 2, a cytokine which controls the production, differentiation, and function of granulocytes and macrophages. The encoded protein is a member of the cytokine family of receptors. This gene is found in the pseudoautosomal region (PAR) of the X and Y chromosomes. Multiple transcript variants encoding different isoforms have been found for this gene, with some of the isoforms being membrane-bound and others being soluble.Specifications
CD116 | |
0.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
CD116; CD116 antigen; CDw116; colony stimulating factor 2 receptor alpha chain; colony stimulating factor 2 receptor alpha subunit; colony stimulating factor 2 receptor, alpha, low-affinity (granulocyte-macrophage); CSF 2R; CSF2R; Csf2ra; CSF2RAX; CSF2RAY; CSF2RX; CSF2RY; Csfgmra; GM CSF R alpha; GM-CSF receptor alpha subunit; GMCSFR; GM-CSFR; GM-CSF-Ra; GM-CSFRalpha; GM-CSF-R-alpha; GMCSFR-alpha; GMR; GMR-alpha; granulocyte-macrophage colony stimulating receptor alpha; granulocyte-macrophage colony-stimulating factor receptor alpha chain; granulocyte-macrophage colony-stimulating factor receptor subunit alpha; MGC3848; MGC4838; SMDP4 | |
CSF2RA | |
Primary | |
4°C | |
CSF2RA |
4H1 | |
Flow Cytometry | |
Unconjugated | |
Mouse | |
Human | |
P15509 | |
1438 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.