Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD116 Mouse anti-Human, BUV395, Clone: hGMCSFR-M1, BD Optibuild™

Catalog No. BDB743382 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB743382 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB743382 Supplier BD Biosciences Supplier No. 743382
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The hGMCSFR-M1 antibody reacts with the subunit (GM-CSFR) of the human Granulocyte-Macrophage Colony-Stimulating Factor Receptor complex. This 75-85 kD subunit is also known as CD116. The hGMCSFR-M1 antibody was first clustered at the Fifth International Workshop on Human Leucocyte Differentiation Antigens. The GM-CSFR subunit associates with the 120-140 kDβc subunit (common subunit, CD131), that is shared with the receptors for interleukins IL-3 and IL-5. Both of the chains of the GM-CSFR complex are involved in ligand binding and intracellular signaling. Theαchain appears to transmit most of the biological signals. CD116 is expressed by a variety of myeloid cell lines, hematopoietic and non-hematopoetic tumor cells, and normal cell types including monocytes, macrophages, neutrophils, eosinophils, myeloid dendritic cells, endothelial cells, fibroblasts, and placental trophoblasts. Lymphocytes are negative for GM-CSFR expression. Reports suggest that GM-CSFR plays a role in myeloid lineage growth and differentiation. The immunogen used to generate the hGMCSFR-M1 hybridoma was recombinant human GM-CSFR.

The antibody was conjugated to BD Horizon™ BUV395 which is part of the BD Horizon Brilliant™ Ultraviolet family of dyes. This dye has been exclusively developed by BD Biosciences to have minimal spillover into other detectors, making it an optimal choice for multicolor flow cytometry. With an Ex Max at 348 nm and an Em Max at 395 nm, BD Horizon BUV395 can be excited with a 355 nm laser and detected with a 379/28 filter. BUV395 is a UV-excitable dye that has been developed exclusively by BD Biosciences. With an excitation max of 348 nm and emission max of 395 nm, BUV395 can be excited by the 355nm laser and detected in a 379/28 filter. This dye is optimal for multicolor flow cytometry because it has little to no spillover into other detectors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD116
Classification Monoclonal
Clone hGMCSFR-M1
Concentration 0.2mg/mL
Conjugate Brilliant Ultraviolet 395
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias CSF2RA, GM-CSF Receptor alpha, GM-CSFRα, GMCSFRA, GMR, SMDP4
Host Species Mouse
Immunogen Recombinant human GM-CSFR
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.