Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD116 Recombinant Rabbit Monoclonal Antibody (JE61-37)
GREENER_CHOICE

Catalog No. FEMA550762
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550762 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550762 Supplier Invitrogen™ Supplier No. MA550762
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive control: JAR cell lysate, Hela cell lysate, 293T cell lysate, human placenta tissue lysate, human malignant melanoma tissue, human skin tissue, SiHa. Predicted band size: 46 kDa Subcellular Location: Cell membrane; Secreted.

The protein encoded by this gene is the alpha subunit of the heterodimeric receptor for colony stimulating factor 2, a cytokine which controls the production, differentiation, and function of granulocytes and macrophages. The encoded protein is a member of the cytokine family of receptors. This gene is found in the pseudoautosomal region (PAR) of the X and Y chromosomes. Multiple transcript variants encoding different isoforms have been found for this gene, with some of the isoforms being membrane-bound and others being soluble.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD116
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone JE61-37
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene CSF2RA
Gene Accession No. P15509
Gene Alias CD116; CD116 antigen; CDw116; colony stimulating factor 2 receptor alpha chain; colony stimulating factor 2 receptor alpha subunit; colony stimulating factor 2 receptor, alpha, low-affinity (granulocyte-macrophage); CSF 2R; CSF2R; Csf2ra; CSF2RAX; CSF2RAY; CSF2RX; CSF2RY; Csfgmra; GM CSF R alpha; GM-CSF receptor alpha subunit; GMCSFR; GM-CSFR; GM-CSF-Ra; GM-CSFRalpha; GM-CSF-R-alpha; GMCSFR-alpha; GMR; GMR-alpha; granulocyte-macrophage colony stimulating receptor alpha; granulocyte-macrophage colony-stimulating factor receptor alpha chain; granulocyte-macrophage colony-stimulating factor receptor subunit alpha; MGC3848; MGC4838; SMDP4
Gene Symbols CSF2RA
Host Species Rabbit
Immunogen Recombinant protein within Human CD116 aa 23-122/400.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1438
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.