Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD117/c-kit Antibody (KIT/982), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311446B
Description
CD117/c-kit Monoclonal antibody specifically detects CD117/c-kit in Human samples. It is validated for Western BlotSpecifications
| CD117/c-kit | |
| Monoclonal | |
| Biotin | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| KIT/982 | |
| Western Blot | |
| CD117, CD117 antigen, ckit, C-Kit, EC 2.7.10, EC 2.7.10.1, PBT, piebald trait, Proto-oncogene c-Kit, proto-oncogene tyrosine-protein kinase Kit, SCFRmast/stem cell growth factor receptor, soluble KIT variant 1, Tyrosine-protein kinase Kit, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene-like protein | |
| Recombinant fragment (around aa100-300) of human CD117/c-kit protein (exact sequence is proprietary) (Uniprot: P10721) | |
| 0.1 mL | |
| B Cell Development and Differentiation Markers, Cancer, Cancer Stem Cells, Cellular Markers, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Mast Cell Markers, Mesenchymal Stem Cell Markers, Myeloid derived Suppressor Cell, Phospho Specific, Protein Kinase, Signal Transduction, Stem Cell Markers, Tyrosine Kinases | |
| 3815 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction