Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD117 (c-Kit) Mouse anti-Mouse, Human, Unconjugated, Clone: 2C11, MilliporeSigma™

Catalog No. MABF2053100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF2053-100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF2053-100 Supplier MilliporeSigma Supplier No. MABF2053100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This mouse monoclonal Anti-CD117 (c-Kit), clone 2C11 Antibody, Cat. No. MABF2053 is tested for use in Western Blotting and Flow Cytometry, for the detection of C-kit.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD117 (c-Kit)
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone 2C11
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Mast/stem cell growth factor receptor Kit;EC: 2.7.10.1;SCFR;Piebald trait protein;PBT;Proto-oncogene c-Kit;Tyrosine-protein kinase Kit;p145 c-kit;v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog
Gene Symbols KIT;SCFR
Host Species Mouse
Immunogen GST-tagged recombinant fragment corresponding to 195 amino acids from the extracellular domain of Human C-kit.
Molecular Weight of Antigen ∽160 kDa observed; 109.87 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Inflammation & Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_000213.1
Test Specificity Clone 2C11 specifically detects CD117 (c-Kit) in human and murine cells. It targets an epitope with in the extracellular domain.
Target Species Mouse, Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.