Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD117 (c-kit) Rat anti-Mouse, BV 421, Clone: ACK2, BD Horizon™
Rat Monoclonal Antibody
Supplier: BD Biosciences 567818
This item is not returnable.
View return policy
Description
CD117 (c-kit) Monoclonal antibody specifically detects CD117 (c-kit) in Mouse samples. It is validated for Flow Cytometry
Specifications
CD117 (c-kit) | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Rat | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. | |
IgG2b κ |
Flow Cytometry | |
ACK2 | |
Brilliant Violet 421 | |
CD117; c-KIT; Kit; SCFR; Steel Factor Receptor | |
Mouse IL-3-dependent Mast Cells | |
RUO | |
Mouse | |
Liquid |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction