Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD119 (IFN gamma Receptor 1) Monoclonal Antibody (2E2), PE, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 501122260
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-112-2260 100 μg
50-105-80 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-112-2260 Supplier Invitrogen™ Supplier No. 12119182
Add to Cart
Edge
Add to Cart

Armenian Hamster Monoclonal Antibody

Description: The monoclonal antibody 2E2 recognizes mouse CD119 also known as IFN gamma receptor1. CD119 heterodimerizes with IFN gamma receptor 2 (AF-1) forming the Class II cytokine receptor. The ligand IFN gamma binds IFN gamma receptor 1 subunit but signal transduction transpires via the IFN gamma receptor 2 subunit. The receptor is involved in host defense and expression is broad including B cells, NK cells, macrophages and dendritic cells as well as other non-hematopoietic cell types. Applications Reported: This 2E2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 2E2 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. In addition, longer incubations appear to help stain the popμLation; therefore we recommend a 60 minute incubation at 2-8°C. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.

This gene (IFNGR1) encodes the ligand-binding chain (alpha) of the gamma interferon receptor. Human interferon-gamma receptor is a heterodimer of IFNGR1 and IFNGR2. A genetic variation in IFNGR1 is associated with susceptibility to Helicobacter pylori infection. In addition, defects in IFNGR1 are a cause of mendelian susceptibility to mycobacterial disease, also known as familial disseminated atypical mycobacterial infection.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD119 (IFN gamma Receptor 1)
Applications Flow Cytometry
Classification Monoclonal
Clone 2E2
Concentration 0.2 mg/mL
Conjugate PE
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene IFNGR1
Gene Accession No. P15261
Gene Alias antiviral protein, type 2; AVP, type 2; CD119; CD119 antigen; CDw119; Ifgr; IFN-gamma receptor 1; IFN-gammaR; IFN-gamma-R1; IFN-gamma-R-alpha; Ifngr; IFNGR1; IMD27A; IMD27B; immune interferon receptor 1; INF-g receptor; interferon gamma receptor 1; Interferon gamma receptor alpha-chain; interferon-gamma receptor 1; interferon-gamma receptor alpha chain; Nktar
Gene Symbols IFNGR1
Host Species Armenian Hamster
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15979
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.