Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD11a/CD18 Antibody (hu1124 (Efalizumab)), Alexa Fluor™ 647, Novus Biologicals™

Human Monoclonal Antibody
Supplier: Novus Biologicals NBP275912AF647
Description
CD11a/CD18 Monoclonal specifically detects CD11a/CD18 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry.Specifications
CD11a/CD18 | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ITGAL | |
Efalizumab binds to CD11a, a subunit of leukocyte function antigen-1 (LFA-1), which is expressed on all leukocytes. | |
0.1 mL | |
Primary | |
Binds to human LFA-1. | |
Store at 4°C in the dark. |
Flow Cytometry, Immunohistochemistry | |
hu1124 (Efalizumab) | |
Flow Cytometry, Immunohistochemistry | |
CD11 antigen-like family member A, CD11a antigen, CD11Aantigen CD11A (p180), lymphocyte function-associated antigen 1, alphapolypeptide, integrin alpha-L, integrin gene promoter, integrin, alpha L (antigen CD11A (p180), lymphocyte function-associated antigen1; alpha polypeptide), Leukocyte adhesion glycoprotein LFA-1 alpha chain, Leukocyte function-associated molecule 1 alpha chain, LFA-1, LFA-1 alpha, LFA1A, LFA-1A, lymphocyte function-associated antigen 1 | |
Human | |
Protein A purified | |
Cytokine Research, Immunology, Signal Transduction | |
3683.0 | |
Human | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction