Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD11a/CD18 Antibody (HUH73A), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP260802
Description
CD11a/CD18 Monoclonal specifically detects CD11a/CD18 in Human, Bovine, Canine, Equine, Feline, Goat, Bison, Sheep samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
CD11a/CD18 | |
Monoclonal | |
1 mg/mL | |
Flow Cytometry 1:10 - 1:1000 | |
CD11 antigen-like family member A, CD11a antigen, CD11Aantigen CD11A (p180), lymphocyte function-associated antigen 1, alphapolypeptide, integrin alpha-L, integrin gene promoter, integrin, alpha L (antigen CD11A (p180), lymphocyte function-associated antigen1; alpha polypeptide), Leukocyte adhesion glycoprotein LFA-1 alpha chain, Leukocyte function-associated molecule 1 alpha chain, LFA-1, LFA-1 alpha, LFA1A, LFA-1A, lymphocyte function-associated antigen 1 | |
Mouse | |
Unpurified | |
Cytokine Research, Immunology, Signal Transduction | |
3683 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Flow Cytometry, Immunohistochemistry, Functional Assay | |
HUH73A | |
Unconjugated | |
Ascites with 0.09% Sodium Azide | |
ITGAL | |
Human mononuclear leukocytes | |
0.1 mg | |
Primary | |
Human, Bovine, Canine, Equine, Feline, Goat, Bison, Sheep | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction