Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD11a/CD18 Monoclonal Antibody (CVS9), PE
GREENER_CHOICE

Catalog No. PIMA528273
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA528273 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA528273 Supplier Invitrogen™ Supplier No. MA528273
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse anti Horse CD11a/CD18 antibody, clone CVS9 recognizes the equine homolog of the human CD11a/CD18 cell surface antigen, a heterodimer expressed on all equine cells of haemopoietic origin. Reconstitute by adding 1 mL distilled water.

CD11/CD18 (LFA-1) are beta 2-type integrins. The leukocyte integrins are a family of heterodimeric receptors that mediate divalent cation-dependent cellular adhesion reactions. T cells use integrins in essentially all of their functions. Integrins become active following signalling through other membrane receptors, which cause both affinity alteration and an increase in integrin clustering. The monoclonal antibody 24 recognizes a structural feature and is strictly dependent upon the presence of Mg2+. The epitope is located within, or in close proximity to, the three conserved cation binding domains and therefore a measure of Mg2+ bound to the leukocyte integrins and thus reflects functionally active molecules. The epitope can be induced on polymorphonuclear leukocytes and on monocytes. Glu173 and Glu175 of the beta(2) I domain are identified as critical for antibody 24 binding. The monoclonal antibody 24 inhibits monocyte-dependent, antigen specific T cell proliferation and IL-2- activated natural killer cell assays. The antibody does not interfere with mitogen-stimulated T cell proliferation. Furthermore the monoclonal antibody 24 prevents deadhesion of receptor/ligand pairs, possible locking leukocyte integrins in an active conformation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD11a/CD18
Applications Flow Cytometry
Classification Monoclonal
Clone CVS9
Concentration Conc. Not Determined
Conjugate PE
Formulation PBS with 1% BSA, 5% sucrose and 0.09% sodium azide
Gene ITGAL
Gene Accession No. 0
Gene Alias antigen CD11A (p180); antigen CD11A (p180), lymphocyte function-associated antigen 1, alpha polypeptide; CD11 antigen-like family member A; CD11a; CD18; cell surface adhesion glycoprotein (LFA-1/CR3/P150,959 beta subunit precursor); Cell surface adhesion glycoproteins LFA-1/CR3/p150,95 subunit beta; complement component 3 receptor 3 and 4 subunit; complement receptor C3 beta-subunit; Complement receptor C3 subunit beta; integrin alpha L; integrin alpha-L; integrin beta chain, beta 2; integrin beta-2; integrin gene promoter; integrin subunit alpha L; integrin subunit beta 2; integrin, alpha L (antigen CD11A (p180), lymphocyte function-associated antigen 1; alpha polypeptide); integrin, beta 2 (complement component 3 receptor 3 and 4 subunit); ITGAL; ITGB2; LAD; LCAMB; Leukocyte adhesion glycoprotein LFA-1 alpha chain; leukocyte cell adhesion molecule CD18; leukocyte function-associated molecule 1 alpha chain; leukocyte-associated antigens CD18/11A, CD18/11B, CD18/11C; LFA-1; LFA-1 alpha; LFA1A; LFA-1A; lymphocyte function-associated antigen 1; lymphocyte function-associated antigen 1, alpha polypeptide; MAC-1; MF17; MFI7
Gene Symbols ITGAL, ITGB2
Host Species Mouse
Immunogen Equine leucocytes.
Purification Method Protein G
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100056128, 100065508
Target Species Horse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.