Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD11a (LFA-1A) Mouse anti-Human, RB545, BD OptiBuild™

Catalog No. BDB756527 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB756527 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB756527 Supplier BD Biosciences Supplier No. 756527
Add to Cart
Add to Cart

Mouse Primary Antibody

CD11a (LFA-1A) Primary antibody specifically detects CD11a (LFA-1A) in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD11a (LFA-1A)
Applications Flow Cytometry
Clone G25.2 (also known as G-25.2)
Concentration 0.2 mg/mL
Conjugate RB545
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias ITGAL; LFA-1 alpha; LFA-1A; LFA1A; integrin alpha L
Host Species Mouse
Immunogen Human Cytotoxic T Lymphocytes
Quantity 50 μg
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.