Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD11a (LFA-1alpha) Monoclonal Antibody (M17/4), NovaFluor™ Red 700, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
$201.00 - $379.00
Specifications
Antigen | CD11a (LFA-1alpha) |
---|---|
Clone | M17/4 |
Concentration | 0.1 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
M054T03R03A
![]() |
Invitrogen™
M054T03R03A |
100 μg |
Each for $379.00
|
|
|||||
M054T02R03A
![]() |
Invitrogen™
M054T02R03A |
25 μg |
Each for $201.00
|
|
|||||
Description
CD11a (LFA-1alpha) Monoclonal antibody specifically detects CD11a (LFA-1alpha) in Mouse samples. It is validated for Flow CytometrySpecifications
CD11a (LFA-1alpha) | |
0.1 mg/mL | |
Monoclonal | |
Rat | |
Mouse | |
P24063 | |
16408 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! |
M17/4 | |
Flow Cytometry | |
NovaFluor Red 700 | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
(p180); alpha L integrin; antigen CD11A (p180); antigen CD11A (p180), lymphocyte function-associated antigen 1, alpha polypeptide; antigen CD11A p180; CD11 antigen-like family member A; CD11a; integrin alpha L; integrin alphaL; integrin alpha-L; integrin alpha-L precursor; integrin gene promoter; integrin subunit alpha L; integrin surface receptor; integrin, alpha L; integrin, alpha L (antigen CD11A (p180), lymphocyte function-associated antigen 1; alpha polypeptide); ITGAL; Leukocyte adhesion glycoprotein LFA-1 alpha chain; leukocyte function-associated molecule 1 alpha chain; LFA-1; LFA-1 alpha; LFA1A; LFA-1A; Ly-15; Ly-21; lymphocyte antigen 15; lymphocyte function associated antigen 1, alpha polypeptide; lymphocyte function-associated antigen 1; lymphocyte function-associated antigen 1, alpha polypeptide; lymphocyte function-associated antigen-1 | |
IgG2a κ | |
Affinity chromatography | |
ITGAL |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title