Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD11a (LFA-1alpha) Monoclonal Antibody (R7.1), eBioscience™
GREENER_CHOICE

Catalog No. 5017682
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-176-82 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-176-82 Supplier Invitrogen™ Supplier No. BMS102
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody specifically recognises CD11a, otherwise known as integrin alpha L chain. Clone R7.1 recognises an epitope within the I-domain of CD11a and can be used in neutralization studies. BMS102 is also suitable as primary antibody in staining for FACS analysis, BMS102FI can be used for direct staining of cells, BMS102BT can be used in a 2-step procedure using the biotin-(strept)avidin system. CD11a is a 180kDa glycoprotein and member of the integrin family of cell adhesion molecules, which is expressed by all human leukocytes.CD11a combines with the CD18 beta chain (ITGB2) to form lymphocyte function-associated antigen-1 (LFA-1), and binds to its major ligands CD50 (ICAM3), CD54 (ICAM1) and CD102 (ICAM2), acting as a key mediator of cell-cell adhesion processes in normal immune and inflammatory responses. Applications Tested: Flow Cytometry, Functional Studies (Blocking), Immunohistochemistry. Purity: >95%.

CD11a (LFA-1 alpha, ITGAL) together with CD18 constitute leukocyte function-associated antigen 1 (LFA-1), the alphaLbeta2 integrin. CD11a is implicated in activation of LFA-1 complex. CD11a plays a central role in leukocyte intercellular adhesion through interactions with its ligands, ICAMs 1-3 (intercellular adhesion molecules 1 through 3), and also functions in lymphocyte costimulatory signaling. CD11a is expressed on the plasma membrane of leukocytes in a low-affinity conformation. Cell stimulation by chemokines or other signals leads to induction the high-affinity conformation, which supports tight binding of CD11a to its ligands, the intercellular adhesion molecules ICAM-1, -2, -3. CD11a is thus involved in interaction of various immune cells and in their tissue-specific settlement, but participates also in control of cell differentiation and proliferation and of T-cell effector functions. Blocking of CD11a function by specific antibodies or small molecules has become an important therapeutic approach in treatment of multiple inflammatory diseases. For example, humanized anti- CD11a antibody Efalizumab (Raptiva) is being used to interfere with T cell migration to sites of inflammation, binding of cholesterol-lowering drug simvastatin to CD11a allosteric site leads to immunomodulation and increase in lymphocytic cholinergic activity. Two transcript variants encoding different isoforms of CD11a have been found. Diseases associated with CD11a dysfunction include benign cephalic histiocytosis and leukocyte adhesion deficiency.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD11a (LFA-1alpha)
Applications Flow Cytometry, Functional Assay, Immunohistochemistry
Classification Monoclonal
Clone R7.1
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation TBS with no preservative; pH 7.2-7.4
Gene ITGAL
Gene Accession No. P20701
Gene Alias (p180); alpha L integrin; antigen CD11A (p180); antigen CD11A (p180), lymphocyte function-associated antigen 1, alpha polypeptide; antigen CD11A p180; CD11 antigen-like family member A; CD11a; integrin alpha L; integrin alphaL; integrin alpha-L; integrin alpha-L precursor; integrin gene promoter; integrin subunit alpha L; integrin surface receptor; integrin, alpha L; integrin, alpha L (antigen CD11A (p180), lymphocyte function-associated antigen 1; alpha polypeptide); ITGAL; Leukocyte adhesion glycoprotein LFA-1 alpha chain; leukocyte function-associated molecule 1 alpha chain; LFA-1; LFA-1 alpha; LFA1A; LFA-1A; Ly-15; Ly-21; lymphocyte antigen 15; lymphocyte function associated antigen 1, alpha polypeptide; lymphocyte function-associated antigen 1; lymphocyte function-associated antigen 1, alpha polypeptide; lymphocyte function-associated antigen-1
Gene Symbols ITGAL
Host Species Mouse
Immunogen Rec. Human CD11a (LFA-1alpha).
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3683
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.