Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD11a, Mouse anti-Human, Clone: BU17, Millipore Sigma™

Catalog No. MABF2003100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF2003100 100 μg
MABF200325 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF2003100 Supplier MilliporeSigma Supplier No. MABF2003100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-CD11a, clone BU17, Cat. No. MABF2003, is a mouse monoclonal antibody that detects CD11a and has been tested for use in Flow Cytometry.

Integrin alpha-L (UniProt: P20701; also known as CD11 antigen-like family member A, Leukocyte adhesion glycoprotein LFA-1 alpha chain, LFA-1A, Leukocyte function-associated molecule 1 alpha chain, CD11a) is encoded by the ITGAL (also known as CD11A) gene (Gene ID: 3683) in human. Integrins are part of a family of heterodimeric transmembrane glycoproteins that recognize a variety of ligands and control numerous physiologic functions in cells. CD11a is a single-pass type I membrane protein that noncovalently associates with beta2 integrin to forms a heterodimer and serve as a receptor for ICAM1, ICAM2, ICAM3 and ICAM4. It is expressed on B and T lymphocytes, monocytes, macrophages, neutrophils, basophils, and eosinophils. It is also involved in generation of common lymphoid progenitor cells in bone marrow. In resting T-cells, about 40% of CD11a is constitutively phosphorylated and phosphorylation causes conformational changes needed for ligand binding and is considered to be essential for activation by some physiological agents. CD11a is synthesized with a signal peptide (aa 1-25), which is subsequently cleaved off in the mature form. It has a large extracellular domain (aa 26-1090), a transmembrane domain (aa 1091-1111), and a cytoplasmic domain 1112-1170). Three isoforms of CD11a have been described that are produced by alternative splicing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD11a
Applications Flow Cytometry
Classification Monoclonal
Clone BU17
Conjugate Unconjugated
Dilution Flow Cytometry Analysis: A representative lot detected CD11a in Flow Cytometry applications (Baker, M.P., et. al. (1998). Blood. 92(8):2830-43).
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Integrin alpha-L;CD11 antigen-like family member A;Leukocyte adhesion glycoprotein LFA-1 alpha chain;LFA-1A;Leukocyte function-associated molecule 1 alpha chain
Gene Symbols ITGAL;CD11A
Host Species Mouse
Immunogen Waldenstrom macroglobulinemia lymphoplasmacytic cells.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Inflammation & Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_002200.2
Test Specificity Clone BU17 detects CD11a in human cells. It targets an epitope with in the extracellular domain.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.