Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD11a Mouse, anti-Human, R718, Clone: G-25.2, BD OptiBuild™

Catalog No. BDB752246 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752246 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752246 Supplier BD Biosciences Supplier No. 752246
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The G-25.2 monoclonal antibody specifically recognizes CD11a which is also known as Leukocyte function-associated molecule 1 alpha chain (LFA-1A or LFA-1α). CD11a is a ∼180 kDa type I transmembrane glycoprotein that is encoded by ITGAL (Integrin subunit alpha L) that belongs to integrin α-chain family. CD11a is a leucocyte adhesion receptor that noncovalently complexes with CD18 to form the LFA-1 heterodimer (CD11a/CD18) which binds to intercellular adhesion molecules (ICAMs) expressed on target tissues including ICAM-1 (CD54), ICAM-2 (CD102), ICAM-3 (CD50), ICAM-4, or ICAM-5. The CD11a antigen is expressed on virtually all normal peripheral blood leucocytes. It is essential for many immune responses that require cell-to-cell contact, such as lymphocyte adhesion, natural killer (NK) and T-lymphocyte cytolysis, and T lymphocyte proliferation. Activation of lymphocytes through the T-cell antigen receptor (TCR) upregulates LFA-1 function, converting the antigen to a high-avidity state. Expression of LFA-1 on monocytes is upregulated by γ-interferon (IFN-γ). While normal cells possess both the α and β LFA-1 chains, cells of subjects with some leukemias, lymphomas, or leucocyte adhesion deficiency (LAD) may lack the entire LFA-1 molecule or the α or β chain. The absence of LFA-1 on tumor cells may provide a mechanism for their escape from immune surveillance.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD11a
Applications Flow Cytometry
Classification Monoclonal
Clone G-25.2
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.