Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD11a Mouse, anti-Rat, R718, Clone: WT.1, BD OptiBuild™

Catalog No. BDB751898 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB751898 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB751898 Supplier BD Biosciences Supplier No. 751898
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The WT.5 monoclonal antibody specifically recognizes the alpha subunit of LFA-1 (αLβ2 integrin, CD11a/CD18), a heterodimeric surface glycoprotein which is found on the majority of leukocytes, but not on peritoneal macrophages or peritoneal mast cells. LFA-1 mediates a variety of heterotypic and homotypic intercellular adhesions through interaction with ICAM-1 (CD54) and ICAM-2 (CD102). WT.1 mAb recognizes both the activated and unactivated forms of LFA-1. It inhibits the binding of LFA-1 to ICAM-1 in several in vitro assays, including binding of Concanavalin A-stimulated lymphocytes (Con A blasts) to purified ICAM-1 and Mg2+-dependent aggregation of concanavalin A-stimulated blasts. It has also been reported to inhibit leukocyte infiltration in several in vivo models of inflammation.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD11a
Applications Flow Cytometry
Classification Monoclonal
Clone WT.1
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Rat
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.