Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD11a Rat anti-Mouse, BB700, Clone: M17/4, BD Optibuild™

Catalog No. BDB742109 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB742109 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB742109 Supplier BD Biosciences Supplier No. 742109
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The M17/4 antibody reacts with the 180 kDa αL chain of LFA-1 (CD11a/CD18, αLβ2 integrin), a heterodimeric surface glycoprotein expressed on almost all leukocytes. CD8a+CD8b- intestinal intraepithelial T lymphocytes, which are believed to be thymus independent, do not express CD11a. LFA-1 mediates a variety of heterotypic and homotypic intercellular adhesions through interaction with ICAM-1 (CD54) and ICAM-2 (CD102), including participation in the immunological synapses between CD8+ T lymphocytes and antigen-presenting cells. mAb M17/4 blocks a variety of LFA-1-mediated cells interactions in vitro, and costimulatory effects have also been described. In vivo treatment with M17/4 mAb reduces the severity of graft-versus-host reactions, prolongs allograft survival, inhibits the development of autoimmunity, and blocks substance P-induced leukocyte migration. The M17/4 and 2D7 (Cat. No. 553120) antibodies are reported to recognize different epitopes of the CD11a molecule.

The antibody was conjugated to BD Horizon™ BB700, which is part of the BD Horizon Brilliant™ Blue family of dyes. It is a polymer-based tandem dye developed exclusively by BD Biosciences. With an excitation max of 485 nm and an emission max of 693 nm, BD Horizon BB700 can be excited by the 488 nm laser and detected in a standard PerCP-Cy™5.5 set (eg, 695/40nm filter). This dye provides a much brighter alternative to PerCP-Cy5.5 with less cross laser excitation off the 405 nm and 355 nm lasers.BD Horizon Brilliant™ Blue 700 (BB700) is a dye that was exclusively developed by BD Biosciences as brighter alternative to PerCP-Cy5.5. This dye also has less cross laser excitation off the 405 nm laser, resulting in less spillover into the violet channels compared to PerCP-Cy5.5.  Due to similar excitation and emission properties, BD Horizon BB700 and PerCP-Cy5.5 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD11a
Classification Monoclonal
Clone M17/4
Concentration 0.2mg/mL
Conjugate Brilliant Blue 700 (BB700)
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Itgal, Integrin alpha-L, Integrin αL, ITAL, LFA-1A, LFA-1α, Ly-15
Host Species Rat
Immunogen C57BL/6 Mouse Splenic Secondary Cytotoxic T Lymphocytes
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.