Learn More
Description
Specifications
Specifications
| Antigen | CD11a |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | M17/4 |
| Conjugate | Brilliant Ultraviolet 563 |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Gene Alias | Itgal; Integrin alpha-L; Integrin αL; ITAL; LFA-1A; LFA-1α; Ly-15 |
| Host Species | Rat |
| Immunogen | C57BL/6 Mouse Splenic Secondary Cytotoxic T Lymphocytes |
| Quantity | 50 μg |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.