Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD11b Antibody - BSA Free, R&D Systems™
SDP

Catalog No. p-5067470 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11089474T 0.025 mL
NB11089474 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB11089474T Supplier R&D Systems Supplier No. NB11089474SS

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody has been used in 67 publications

CD11b Polyclonal specifically detects CD11b in Bovine, C. elegans, Caenorhabditis elegans, Chinese Hamster, Cricetulus (Chinese Hamster), Human, Monkey, Mouse, Rat, Rhesus Macaque samples. It is validated for Dual RNAScope ISH-IHC, Flow Cytometry, Flow Cytometry (Cell Surface), Immunocytochemistry/Immunofluorescence, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), In-situ Hybridization, Simple Western, Single Cell Western, Western Blot.

Specifications

Antigen CD11b
Applications Dual RNAScope ISH-IHC, Flow Cytometry, Flow Cytometry (Cell Surface), Immunocytochemistry/Immunofluorescence, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), In Situ Hybridization (ISH), Simple Western, Single Cell Western, Western Blot
Classification Polyclonal
Concentration 1.00 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL. Use reported in scientific literature (PMID 31082627), Simple Western 1:50, Flow Cytometry reported in scientific literature (PMID 21422470), Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence 1:200. Use reported in multiple pieces of scientific literature (PMID 23980916), Immunohistochemistry-Paraffin 1:400. Use reported in scientific literature (PMID 31022918), Immunohistochemistry-Frozen reported in scientific literature (PMID 23980916), In-situ Hybridization reported in scientific literature (PMID 27133471), Flow (Cell Surface) 1:10 - 1:1000, Single Cell Western 1:10, Dual RNAscope ISH-IHC
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. P11215, P05555
Gene Alias antigen CD11b (p170), CD11 antigen-like family member B, CD11b, CD11b antigen, Cell surface glycoprotein MAC-1 subunit alpha, Complement component 3 receptor 3 subunit, CR-3 alpha chain, CR3A, CR3AMGC117044, integrin alpha-M, integrin, alpha M (complement
Gene Symbols ITGAM
Host Species Rabbit
Immunogen Rabbit Polyclonal CD11b Antibody was made to a synthetic peptide within residues 250-350 of the mouse CD11b protein. [Swiss-Prot# P05555]
Molecular Weight of Antigen 127.2 kDa
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cellular Markers, Cytoskeleton Markers, Glia Markers, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Microglia Markers, Myeloid Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3684
Test Specificity Mouse, rat and human. We have feedback that this antibody does not work in human samples with Western blot. Immunogen has 100% homology to bovine and 88% homology with sheep.
Target Species Bovine, C. elegans, C. elegans, Chinese Hamster, Chinese Hamster, Human, Monkey, Mouse, Rat, Rhesus Macaque
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Product Type Antibody
Isotype IgG
Product Line Novus
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.