Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD11b Antibody (M1/70.15), PE/Cy7, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NB6001327PECY7
Description
CD11b Monoclonal specifically detects CD11b in Human, Mouse, Rabbit samples. It is validated for Flow Cytometry.Specifications
CD11b | |
Monoclonal | |
PE-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
ITGAM | |
Rat Monoclonal CD11b Antibody (M1/70.15) was made to T cell enriched splenocytes from B10 mice. | |
Protein G purified | |
Breast Cancer, Cancer, Cellular Markers, Cytoskeleton Markers, Glia Markers, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Microglia Markers, Myeloid Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers | |
3684.0 | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
M1/70.15 | |
Flow Cytometry | |
antigen CD11b (p170), CD11 antigen-like family member B, CD11b, CD11b antigen, Cell surface glycoprotein MAC-1 subunit alpha, Complement component 3 receptor 3 subunit, CR-3 alpha chain, CR3A, CR3AMGC117044, integrin alpha-M, integrin, alpha M (complement | |
Rat | |
127.2 kDa | |
0.1 mL | |
Primary | |
Human, Mouse, Rabbit | |
IgG2b λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction