Learn More
Invitrogen™ CD11b/c Monoclonal Antibody (OX-42), Biotin
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA180232
Description
For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Rat CD11b, clone OX-42, recognizes rat CD11b, also known as integrin alpha-M, the receptor for the iC3b component of complement. CD11b is a 1151 amino acid single pass type 1 transmembrane glycoprotein posessing a single vWFA domain and multiple FG-GAP repeats.
Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. CD11b is implicated in various adhesive interactions of monocytes, macrophages and granulocytes as well as in mediating the uptake of complement coated particles. It is identical to CR3, the receptor for the iC3b fragment of the third complement component. CD11b is also a receptor for fibrogen, factor X and ICAM1. It recognizes P1 and P2 peptides of fibrogen gamma chain. The Mac1 CD11b antigen is present on macrophages, granulocytes, natural killer cells, blood monocytes. CD11b is expressed on 8% spleen cells, 44% bone marrow cells and less than 1% thymocytes and is commonly used as a microgial marker in nervous tissue. CD11c encodes the integrin alpha X chain protein. This protein combines with the beta 2 chain (ITGB2) to form a leukocyte-specific integrin referred to as inactivated-C3b (iC3b) receptor 4 (CR4). The alpha X beta 2 complex seems to overlap the properties of the alpha M beta 2 integrin in the adherence of neutrophils and monocytes to stimulated endothelium cells, and in the phagocytosis of complement coated particles. Mutations affecting the CD11b/c gene lead to systemic lupus erythematosus 6.
Specifications
CD11b/c | |
Monoclonal | |
0.1 mg/mL | |
PBS with 1% BSA and 0.09% sodium azide; pH 7.2 | |
0 | |
Itgam, Itgax | |
Rat peritoneal macrophages. | |
100 μg | |
Primary | |
Rat | |
Antibody | |
IgG2a |
Flow Cytometry, Immunohistochemistry (Frozen) | |
OX-42 | |
Biotin | |
Itgam | |
AI449405; antigen CD11b (p170); CD11 antigen-like family member B; CD11 antigen-like family member C; CD11b; CD11B (p170); CD11b/CD18; CD11bc; CD11C; CD11C (p150) alpha polypeptide; cell surface glycoprotein MAC-1 alpha subunit; cell surface glycoprotein MAC-1 subunit alpha; complement component 3 receptor 3 subunit; complement component 3 receptor 4 subunit; complement component receptor 3 alpha-a; complement receptor 4; complement receptor type 3; CR3; CR-3 alpha chain; CR3A; Cr4; F730045J24Rik; integrin alpha M; integrin alpha X; integrin alpha-M; Integrin alpha-X; integrin aX; integrin subunit alpha M; integrin subunit alpha X; integrin, alpha M; integrin, alpha M (complement component 3 receptor 3 subunit); integrin, alpha X; integrin, alpha X (antigen CD11C (p150), alpha polypeptide); integrin, alpha X (complement component 3 receptor 4 subunit); ITGAM; ITGAX; Leu M5; leu M5, alpha subunit; leukocyte adhesion glycoprotein p150,95 alpha chain; Leukocyte adhesion receptor MO1; leukocyte adhesion receptor p150,95; leukocyte surface antigen p150,95, alpha subunit; Ly-40; MAC1; MAC-1; Mac-1 alpha; MAC1A; Mac-1a; macrophage antigen alpha; macrophage antigen alpha polypeptide; MGC117044; MO1A; myeloid membrane antigen, alpha subunit; N418; Neutrophil adherence receptor; neutrophil adherence receptor alpha-M subunit; OX42; OX-42; p150 95 integrin alpha chain; RGD1561123; SLEB6 | |
Mouse | |
Protein A | |
RUO | |
25021, 499271 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.