Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD11b/c Monoclonal Antibody (OX-42), PE
GREENER_CHOICE

Catalog No. PIMA180775
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA180775 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA180775 Supplier Invitrogen™ Supplier No. MA180775
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4°C in the dark. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Rat CD11b, clone OX-42, recognizes rat CD11b, also known as integrin alpha-M, the receptor for the iC3b component of complement. CD11b is a 1151 amino acid single pass type 1 transmembrane glycoprotein posessing a single vWFA domain and multiple FG-GAP repeats.

Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. CD11b is implicated in various adhesive interactions of monocytes, macrophages and granulocytes as well as in mediating the uptake of complement coated particles. It is identical to CR3, the receptor for the iC3b fragment of the third complement component. CD11b is also a receptor for fibrogen, factor X and ICAM1. It recognizes P1 and P2 peptides of fibrogen gamma chain. The Mac1 CD11b antigen is present on macrophages, granulocytes, natural killer cells, blood monocytes. CD11b is expressed on 8% spleen cells, 44% bone marrow cells and less than 1% thymocytes and is commonly used as a microgial marker in nervous tissue. CD11c encodes the integrin alpha X chain protein. This protein combines with the beta 2 chain (ITGB2) to form a leukocyte-specific integrin referred to as inactivated-C3b (iC3b) receptor 4 (CR4). The alpha X beta 2 complex seems to overlap the properties of the alpha M beta 2 integrin in the adherence of neutrophils and monocytes to stimulated endothelium cells, and in the phagocytosis of complement coated particles. Mutations affecting the CD11b/c gene lead to systemic lupus erythematosus 6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD11b/c
Applications Flow Cytometry
Classification Monoclonal
Clone OX-42
Concentration Conc. Not Determined
Conjugate PE
Formulation PBS with 1% BSA, 5% sucrose and 0.09% sodium azide; pH 7.4
Gene Itgam
Gene Accession No. 0
Gene Alias AI449405; antigen CD11b (p170); CD11 antigen-like family member B; CD11 antigen-like family member C; CD11b; CD11B (p170); CD11b/CD18; CD11bc; CD11C; CD11C (p150) alpha polypeptide; cell surface glycoprotein MAC-1 alpha subunit; cell surface glycoprotein MAC-1 subunit alpha; complement component 3 receptor 3 subunit; complement component 3 receptor 4 subunit; complement component receptor 3 alpha-a; complement receptor 4; complement receptor type 3; CR3; CR-3 alpha chain; CR3A; Cr4; F730045J24Rik; integrin alpha M; integrin alpha X; integrin alpha-M; Integrin alpha-X; integrin aX; integrin subunit alpha M; integrin subunit alpha X; integrin, alpha M; integrin, alpha M (complement component 3 receptor 3 subunit); integrin, alpha X; integrin, alpha X (antigen CD11C (p150), alpha polypeptide); integrin, alpha X (complement component 3 receptor 4 subunit); ITGAM; ITGAX; Leu M5; leu M5, alpha subunit; leukocyte adhesion glycoprotein p150,95 alpha chain; Leukocyte adhesion receptor MO1; leukocyte adhesion receptor p150,95; leukocyte surface antigen p150,95, alpha subunit; Ly-40; MAC1; MAC-1; Mac-1 alpha; MAC1A; Mac-1a; macrophage antigen alpha; macrophage antigen alpha polypeptide; MGC117044; MO1A; myeloid membrane antigen, alpha subunit; N418; Neutrophil adherence receptor; neutrophil adherence receptor alpha-M subunit; OX42; OX-42; p150 95 integrin alpha chain; RGD1561123; SLEB6
Gene Symbols Itgam, Itgax
Host Species Mouse
Immunogen Rat peritoneal macrophages.
Purification Method Protein A
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25021, 499271
Target Species Rat
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.