Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD11b/CD18, Mouse anti-Human, Non-human Primate, Clone: mAb107, Millipore Sigma™

Catalog No. MABF208525 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF208525 25 μg
MABF2085100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF208525 Supplier MilliporeSigma Supplier No. MABF208525UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-CD11b/CD18, clone mAb107, Cat. No. MABF2085, is a mouse monoclonal antibody that detects CD11b/CD18 nd has been tested for use in Dot Blot, Flow Cytometry, Immunohistochemistry, and Inhibition assay.

Integrin alpha-M (UniProt: P11215; also known as CD11 antigen-like family member B, CR-3 alpha chain, Cell surface glycoprotein MAC-1 subunit alpha, Leukocyte adhesion receptor MO1, Neutrophil adherence receptor, CD11b) is encoded by the ITGAM (also known as CD11B, CR3A) gene (Gene ID: 3684) in human. Integrins are alpha/beta heterodimeric adhesion receptors that couple the extracellular matrix on other cells with the contractile cytoskeleton to transduce mechanochemical signals across the plasma membrane. CD11b is a single-pass type I membrane protein that is synthesized with a signal peptide (aa 1-16), which is cleaved off in the mature form. It is predominantly expressed in monocytes and granulocytes and some expression is also observed in neutrophils. It is usually found as a heterodimer of alpha and beta subunits and associates with Integrin beta-2 (ITGB2). Two isoforms of CD11b have been reported that are produced by alternative splicing. CD11b contains a large extracellular domain (aa 17-1104), a short transmembrane domain (aa 1105-1128), and a cytoplasmic domain (aa 1129-1152). It has been implicated in various adhesive interactions of monocytes, macrophages and granulocytes as well as in mediating the uptake of complement-coated particles. It contains a metal ion-dependent adhesion site (MIDAS) in the integrin A domain, which is essential for stabilization of domain A in high affinity state. The major ligand recognition site of CD11b/CD18 has been mapped to the A-domain in the CD11b subunit (CD11bA). Both magnesium and calcium ions exhibit equal affinities for the inactive CD11bA, however, upon activation there is a 10-fold increase in the binding affinity of magnesium ions without affecting affinity of calcium ions. Mutations in ITGAM gene are known to cause Systemic lupus erythematosus 6, which is characterized by wide range of system dysfunctions, principally involving skin, joints, kidneys and serosal membranes. (Ref.: Zerria, K., et al. (2006). Immunology 119(4); 431-440; Mahalingam, B., et al., (2011). J. Immunol. 187 (12); 6393-6401; Li, R., et al. (2002). J. Immunol. 168 (3); 1219-1225.).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD11b/CD18
Applications Dot Blot, Flow Cytometry, Immunohistochemistry, Inhibition Assays
Classification Monoclonal
Clone mAb107
Conjugate Unconjugated
Dilution Dot Blot Analysis: A representative lot detected CD11b/CD18 in Dot Blot applications (Li, R., et. al. (2002). J Immunol. 168(3):1219-25). Flow Cytometry Analysis: A representative lot detected CD11b/CD18 in Flow Cytometry applications (Mahalingam, B., et. al. (2011). J Immunol. 187(12):6393-401; Li, R., et. al. (2002). J Immunol. 168(3):1219-25).Inhibition Analaysis: A representative lot prophylactically inactivated CD11b/CD18 and prevents CD11b/CD18 binding to multiple ligands (Dehnadi, A., et. al. (2017). Nat Commun. 8:13899; Li, R., et. al. (2002). J Immunol. 168(3):1219-25). Immunohistochemistry Analysis: A representative lot detected CD11b/CD18 in Immunohistochemistry applications (Dehnadi, A., et. al. (2017). Nat Commun. 8:13899).
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Integrin alpha-M/ITGB2;CR3
Gene Symbols ITGAM;CD11B;CR3A
Host Species Mouse
Immunogen GST-tagged recombinant fragment corresponding to 211 amino acids from the N-terminal half of human CD11bA.
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Gene ID (Entrez) NP_000623
Test Specificity Clone mAb 107 detects CD11b/CD18 in human neutrophils. It targets an epitope with in the A domain.
Target Species Human, Non-human Primate
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.