Learn More
CD11b/Mac-1 Mouse anti-Human, PE-Cy7, Clone: ICRF44, BD
Mouse Monoclonal Antibody
$182.00 - $366.00
Specifications
Antigen | CD11b |
---|---|
Clone | ICRF44 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | PE-Cyanine7 |
Description
The ICRF44 monoclonal antibody specifically binds to CD11b, the 165-kDa adhesion glycoprotein that associates with the 95-kDa integrin β2 (CD18) to form the CD11b/CD18 complex, also known as Mac-1 or CR3. CD11b is expressed on activated lymphocytes, monocytes, granulocytes, and a subset of NK cells. CD11b functions in cell-cell and cell-substrate interactions and is a receptor for iC3b, CD54 (ICAM-1), CD102 (ICAM-2) and CD50 (ICAM-3). This antibody significantly inhibits polymorphonuclear leukocyte aggregation in response to fMLP.
Specifications
CD11b | |
Flow Cytometry | |
PE-Cyanine7 | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
IgG1 κ | |
Affinity Purified | |
Mac-1α, integrin αM subunit, CR3 α chain |
ICRF44 | |
Monoclonal | |
Mouse | |
Human | |
Human monocytes | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.